Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PeproTech MIP-1 gamma Polyclonal Antibody, PeproTech®, Invitrogen™

Catalog No. 500P117100
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
500P117100 100 μg
500P11750UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 500P117100 Supplier PeproTech Supplier No. 500P117100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AA Sequence of recombinant protein: QITHATETKE VQSSLKAQQG LEIEMFHMGF QDSSDCCLSY NSRIQCSRFI GYFPTSGGCT RPGIIFISKR GFQVCANPSD RRVQRCIERL EQNSQPRTYK Q. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Murine MIP-1gamma (CCL9/10). Anti-Murine MIP-1gamma (CCL9/10)-specific antibody was purified by affinity chromatography employing an immobilized Murine MIP-1gamma (CCL9/10)matrix. Sandwich ELISA: To detect Murine MIP-1gamma (CCL9/10) by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Murine MIP-1gamma (CCL9/10) (500-P117Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Murine MIP-1gamma (CCL9/10). Western Blot: To detect Murine MIP-1gamma (CCL9/10) by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. When used in conjunction with compatible secondary reagents, the detection limit for Recombinant Murine MIP-1gamma (CCL9/10) is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

MIP-1gamma is a CC chemokine found in murine blood and a wide variety of murine tissues, with no known human homolog. MIP-1gamma signals through the CCR1 receptor. MIP-1gamma chemoattracts neutrophils, and also inhibits colony formation of bone marrow myeloid immature progenitors. MIP-1gamma contains six cysteines, including the four highly conserved cysteine residues present in CC chemokines.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MIP-1 gamma
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.1-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Ccl9
Gene Accession No. P51670
Gene Alias C-C motif chemokine 9; CCF18; Ccl9; CCL9(29-101); CCL9(30-101); CCL9(31-101); chemokine (C-C motif) ligand 9; chemokine CCL9; macrophage inflammatory protein 1-gamma; Macrophage inflammatory protein-related protein 2; MIP-1 gamma; MIP-1-gamma; Mrp2; MRP-2; RP23-320E6.3; Scya10; Scya9; small inducible cytokine A10; small inducible cytokine A9; Small-inducible cytokine A9
Gene Symbols Ccl9
Host Species Rabbit
Immunogen E.coli-derived Recombinant Murine MIP-1gamma (CCL9/10).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20308
Target Species Mouse
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.