Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MIP-3 alpha Monoclonal Antibody (3N2D9), Rockland™
SDP

Catalog No. 501991351
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1351 25 μL
50-199-1350 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-1351 Supplier Rockland Immunochemicals Supplier No. 209301B51S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This antibody is specific for human MIP-3 alpha protein. A BLAST analysis was used to suggest cross-reactivity with MIP-3 alpha from human sources based on 100% homology with the immunizing sequence. Cross-reactivity with MIP-3 alpha from other sources has not been determined.

Macrophage Inflammatory Protein-3 alpha (MIP-3alpha), also called CCL20, is expressed in the liver, lungs, lymph nodes and peripheral blood lymphocytes. MIP-3alpha is strongly up regulated by inflammatory signals, and down regulated by the anti-inflammatory cytokine IL-10. MIP3alpha signals through the G protein-coupled receptor, CCR6, and acts as a chemoattractant to lymphocytes and dendritic cells.

Specifications

Antigen MIP-3 alpha
Applications Western Blot
Classification Monoclonal
Clone 3N2D9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene CCL20
Gene Accession No. P78556
Gene Alias beta chemokine exodus-1; Beta-chemokine exodus-1; CC chemokine LARC; CC chemokine ST38; C-C motif chemokine 20; C-C motif chemokine ligand 20; Ccl20; CCL20(1-64); CCL20(1-67); CCL20(2-70); chemokine (C-C motif) ligand 20; CKb4; Exodus; exodus-1; Larc; Liver and activation-regulated chemokine; Macrophage inflammatory protein 3 alpha; MIP 3 a; MIP 3 a; MIP 3alpha; MIP 3a; MIP3 a; MIP3 alpha; MIP3 a; MIP-3[a]; MIP3A; MIP-3a; MIP-3-alpha; M-MIP-3-alpha; SCYA20; small inducible cytokine subfamily A (Cys-Cys), member 20; small inducible cytokine subfamily A20; Small-inducible cytokine A20; St38
Gene Symbols CCL20
Host Species Mouse
Immunogen Anti-MIP-3alpha (MOUSE) Monoclonal Antibody was produced in mouse by repeated immunizations with mature full length recombinant human MIP-3alpha produced in E.coli followed by hybridoma development.
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6364
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.