Learn More
Invitrogen™ MIP-3 alpha Monoclonal Antibody (67310)

Description
In ELISAs, this antibody does not cross-react with recombinant human (rh) CCL19, rhCXCL1, rhCCL16, rhCCL11, recombinant mouse CCL20, or recombinant rat CCL20. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
Specifications
Specifications
| Antigen | MIP-3 alpha |
| Applications | ELISA, Neutralization, Western Blot |
| Classification | Monoclonal |
| Clone | 67310 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | CCL20 |
| Gene Accession No. | P78556 |
| Gene Alias | beta chemokine exodus-1; beta-chemokine exodus-1; CC chemokine LARC; CC chemokine ST38; C-C motif chemokine 20; C-C motif chemokine ligand 20; Ccl20; CCL20(1-64); CCL20(1-67); CCL20(2-70); chemokine (C-C motif) ligand 20; CKb4; Exodus; exodus-1; LARC; Liver and activation-regulated chemokine; macrophage inflammatory protein 3 alpha; MIP 3 a; MIP 3 α; MIP 3alpha; MIP 3α; MIP3 a; MIP3 alpha; MIP3 α; MIP-3[a]; MIP3A; MIP-3A; MIP-3-alpha; M-MIP-3-alpha; SCYA20; small inducible cytokine subfamily A (Cys-Cys), member 20; small inducible cytokine subfamily A20; small-inducible cytokine A20; St38 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.