Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PeproTech MIP-3 beta Polyclonal Antibody, PeproTech®, Invitrogen™

Catalog No. 500P95B100
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
50 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
500P95B100 100 μg
500P95B50UG 50 μg
500P95B1MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 500P95B100 Supplier PeproTech Supplier No. 500P95B100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AA Sequence of recombinant protein: GTNDAEDCCL SVTQKPIPGY IVRNFHYLLI KDGCRVPAVV FTTLRGRQLC APPDQPWVER IIQRLQRTSA KMKRRSS. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human MIP-3beta (CCL19). Anti-Human MIP-3beta (CCL19)-specific antibody was purified by affinity chromatography employing an immobilized Human MIP-3beta (CCL19) matrix. Sandwich ELISA: To detect Human MIP-3beta (CCL19) by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human MIP-3beta (CCL19) (500-P95BBt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human MIP-3beta (CCL19). Western Blot: To detect Human MIP-3beta by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human MIP-3beta is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

CCL19, also known as MIP-3 beta (Macrophage Inflammatory Protein 3 beta), is a member of the CC- subfamily of chemokines. It is most closely related to CCL21, with which it shares 32% amino acid sequence homology. CCL19 and CCL21 are expressed mainly by stromal cells in the T cell-rich zones of lymph nodes. They are critical mediators of the homeostatic trafficking of naive T cells and activated dendritic cells into the secondary lymphoid organs. CCL19 and CCL21 also play a role in T cell priming and activation, as well as the recruitment of lymphocytes to inflamed tissue. Both proteins signal via the G protein-coupled receptor, CCR7, which is expressed on T cells and mature dendritic cells. Although CCL19 and CCL21 both exhibit the same affinity for CCR7, only the binding of CCL19 results in the desensitization and internalization of the receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MIP-3 beta
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.1-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Ccl19
Gene Accession No. Q99731
Gene Alias beta chemokine exodus-3; beta-chemokine exodus-3; CC chemokine ligand 19; C-C motif chemokine; C-C motif chemokine 19; C-C motif chemokine ligand 19; Ccl19; chemokine (C-C motif) ligand 19; chemokine CCL19; CK beta-11; CKb11; EBI1 ligand chemokine; EBI-1 ligand chemokine; EBI1-ligand chemokine; EBV-induced molecule 1 ligand chemokine; ELC; Epstein-Barr virus-induced molecule 1 ligand chemokine; exodus-3; macrophage inflammatory protein 3 beta; macrophage inflammatory protein 3-beta; MIP3B; MIP-3b; MIP-3-beta; M-MIP-3-beta; SCYA19; small inducible cytokine A19; small inducible cytokine subfamily A (Cys-Cys), member 19; small-inducible cytokine A19
Gene Symbols Ccl19
Host Species Rabbit
Immunogen E.coli-derived Recombinant Human MIP-3beta (CCL19).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6363
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.