Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MIP-4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119382
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119382 200 μL
PIPA5121889 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5119382 Supplier Invitrogen™ Supplier No. PA5119382
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MIP-4 Polyclonal Antibody for Western Blot, IHC (P)

Macrophage Inflammatory Protein-4 (MIP-4), also called CCL18, is a chemokine expressed in the lymph nodes, lungs, placenta and bone marrow. Although the MIP-4s receptor remains undetermined, MIP-4 is known to act as a chemoattractant for activated and non-activated T cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MIP-4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene CCL18
Gene Accession No. P55774
Gene Alias Alternative macrophage activation-associated CC chemokine 1; AMAC1; AMAC-1; CC chemokine ligand 18; CC chemokine PARC; C-C motif chemokine 18; C-C motif chemokine ligand 18; CCL18; CCL18(1-68); CCL18(3-69); CCL18(4-69); chemokine (C-C motif) ligand 18 (pulmonary and activation-regulated); chemokine (C-C), dendritic; CKb7; CUL9; CUL-9; cullin 9; cullin-9; DCCK1; DC-CK1; dendritic cell chemokine 1; H7AP1; H-MIP-4; KIAA0708; Macrophage inflammatory protein 4; MIP4; MIP-4; p53-associated parkin-like cytoplasmic protein; PARC; parkin-like cytoplasmic p53 binding protein; pulmonary and activation-regulated chemokine; SCYA18; small inducible cytokine A18; small inducible cytokine subfamily A (Cys-Cys), member 18, pulmonary and activation-regulated; small-inducible cytokine A18; UbcH7-associated protein 1
Gene Symbols CCL18
Host Species Rabbit
Immunogen Recombinant protein Pulmonary Activation Regulated Chemokine. The antigen corresponds to amino acid range 21-89 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6362
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.