Learn More
MIP-4 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
$250.00 - $2175.00
Specifications
Antigen | MIP-4 |
---|---|
Concentration | 0.1-1.0 mg/mL |
Applications | ELISA, Western Blot |
Classification | Polyclonal |
Conjugate | Biotin |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
500P108BT25
![]() |
Invitrogen™
500P108BT25UG |
25 μg |
Each for $250.00
|
|
|||||
500P108BT50
![]() |
Invitrogen™
500P108BT50UG |
50 μg |
Each for $319.00
|
|
|||||
50-271-8709
![]() |
Invitrogen™
500P108BT500UG |
500 μg |
Each for $2,175.00
|
|
|||||
Description
AA Sequence of recombinant protein: QVGTNKELCC LVYTSWQIPQ KFIVDYSETS PQCPKPGVIL LTKRGRQICA DPNKKWVQKY ISDLKLNA. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human MIP-4 (CCL18). Anti-Human MIP-4 (CCL8)-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Human MIP-4 (CCL18) by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human MIP-4 (CCL18) (500-P108) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human MIP-4 (CCL18). Western Blot: To detect hMIP-4 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hMIP-4 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P108BT-1MG will be provided as 2 x 500 μg
Macrophage Inflammatory Protein-4 (MIP-4), also called CCL18, is a chemokine expressed in the lymph nodes, lungs, placenta and bone marrow. Although the MIP-4s receptor remains undetermined, MIP-4 is known to act as a chemoattractant for activated and non-activated T cells.Specifications
MIP-4 | |
ELISA, Western Blot | |
Biotin | |
Rabbit | |
Human | |
P55774 | |
6362 | |
E.coli-derived, 7.8 kDa Recombinant Human MIP-4 (CCL18). | |
Antigen affinity chromatography | |
CCL18 |
0.1-1.0 mg/mL | |
Polyclonal | |
Lyophilized | |
RUO | |
PBS with no preservative | |
Alternative macrophage activation-associated CC chemokine 1; AMAC1; AMAC-1; CC chemokine ligand 18; CC chemokine PARC; C-C motif chemokine 18; C-C motif chemokine ligand 18; CCL18; CCL18(1-68); CCL18(3-69); CCL18(4-69); chemokine (C-C motif) ligand 18 (pulmonary and activation-regulated); chemokine (C-C), dendritic; CKb7; CUL9; CUL-9; cullin 9; cullin-9; DCCK1; DC-CK1; dendritic cell chemokine 1; H7AP1; H-MIP-4; KIAA0708; Macrophage inflammatory protein 4; MIP4; MIP-4; p53-associated parkin-like cytoplasmic protein; PARC; parkin-like cytoplasmic p53 binding protein; pulmonary and activation-regulated chemokine; SCYA18; small inducible cytokine A18; small inducible cytokine subfamily A (Cys-Cys), member 18, pulmonary and activation-regulated; small-inducible cytokine A18; UbcH7-associated protein 1 | |
CCL18 | |
Primary | |
-20°C |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.