Learn More
Invitrogen™ MIP-4 Recombinant Rabbit Monoclonal Antibody (016)

Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human CCL18/PARC/MIP-4.
Specifications
Specifications
| Antigen | MIP-4 |
| Applications | ELISA |
| Classification | Recombinant Monoclonal |
| Clone | 016 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | CCL18 |
| Gene Accession No. | P55774 |
| Gene Alias | Alternative macrophage activation-associated CC chemokine 1; AMAC1; AMAC-1; CC chemokine ligand 18; CC chemokine PARC; C-C motif chemokine 18; C-C motif chemokine ligand 18; CCL18; CCL18(1-68); CCL18(3-69); CCL18(4-69); chemokine (C-C motif) ligand 18 (pulmonary and activation-regulated); chemokine (C-C), dendritic; CKb7; CUL9; CUL-9; cullin 9; cullin-9; DCCK1; DC-CK1; dendritic cell chemokine 1; H7AP1; H-MIP-4; KIAA0708; Macrophage inflammatory protein 4; MIP4; MIP-4; p53-associated parkin-like cytoplasmic protein; PARC; parkin-like cytoplasmic p53 binding protein; pulmonary and activation-regulated chemokine; SCYA18; small inducible cytokine A18; small inducible cytokine subfamily A (Cys-Cys), member 18, pulmonary and activation-regulated; small-inducible cytokine A18; UbcH7-associated protein 1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.