Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MIPEP Antibody (OTI5E7), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272706PE
Description
MIPEP Monoclonal specifically detects MIPEP in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
MIPEP | |
Monoclonal | |
PE | |
PBS with 0.05% Sodium Azide | |
MIPEP | |
Recombinant protein fragment corresponding to amino acids 174-516 of human MIPEP (NP_005923) produced in E.coli. | |
0.1 mL | |
4285.0 | |
Store at 4°C in the dark. |
Flow Cytometry | |
OTI5E7 | |
Flow Cytometry | |
HMIP, MIPEC 3.4.24.59, mitochondrial intermediate peptidase | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction