Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MIPEP Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA528269

Catalog No. PIPA528269


Only null left
Add to Cart

Description

Description

Recommended positive controls: A431, HepG2. Predicted reactivity: Mouse (83%), Rat (83%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The product of this gene performs the final step in processing a specific class of nuclear-encoded proteins targeted to the mitochondrial matrix or inner membrane. This protein is primarily involved in the maturation of oxidative phosphorylation (OXPHOS)-related proteins. This gene may contribute to the functional effects of frataxin deficiency and the clinical manifestations of Friedreich ataxia.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MIPEP
Polyclonal
Unconjugated
MIPEP
HMIP; MIP; MIPEP; Mitochondrial intermediate peptidase
Rabbit
Antigen affinity chromatography
RUO
4285
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.53 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q99797
MIPEP
Recombinant protein encompassing a sequence within the center region of human MIPEP. The exact sequence is proprietary.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.