Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MIPEP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109959
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109959 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109959 Supplier Invitrogen™ Supplier No. PA5109959
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QHVTGTRCPT DFAEVPSILM EYFANDYRVV NQFARHYQTG QPLPKNMVSR LCESKKVCAA ADMQLQVFYA TLDQIYHGKH PLRNSTTDIL KETQEKFYGL PYVPNTAWQL RFSHLVGYGA RYYSYLMSRA VASMVWKECF LQDPFNRAAG ERYRREMLAH GGGREPMLMV EGMLQKCPSV DDFVSALVSD LDLDFETFLM DSE.

The product of this gene performs the final step in processing a specific class of nuclear-encoded proteins targeted to the mitochondrial matrix or inner membrane. This protein is primarily involved in the maturation of oxidative phosphorylation (OXPHOS)-related proteins. This gene may contribute to the functional effects of frataxin deficiency and the clinical manifestations of Friedreich ataxia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MIPEP
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.74 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene MIPEP
Gene Accession No. Q99797
Gene Alias HMIP; MIP; MIPEP; Mitochondrial intermediate peptidase
Gene Symbols MIPEP
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 511-713 of human MIPEP (NP_0059232).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4285
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.