Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MIPOL1 Monoclonal Antibody (1B7)
GREENER_CHOICE

Catalog No. PIMA523072
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA523072 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA523072 Supplier Invitrogen™ Supplier No. MA523072
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: ARETAVQQYK KLEEEIQTLR VYYSLHKSLS QEENLKDQFN YTLSTYEEAL KNRENIVSIT QQQNEELATQ LQQALTERAN MELQLQHARE ASQVANEKVQ KLE.

This gene encodes a coiled-coil domain-containing protein. The encoded protein may function as a tumor suppressor. A translocation that results in truncation of the protein encoded by this locus has been associated with mirror-image polydactyly, also known as Laurin-Sandrow Syndrome. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MIPOL1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 1B7
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene MIPOL1
Gene Accession No. Q8TD10
Gene Alias 1700081O04Rik; 6030439O22Rik; AU021838; AW536152; CCDC193; CDC91L1; D12Ertd19e; MIPOL1; mirror-image polydactyly 1; mirror-image polydactyly gene 1 homolog (human); mirror-image polydactyly gene 1 protein; mirror-image polydactyly gene 1 protein homolog
Gene Symbols MIPOL1
Host Species Mouse
Immunogen MIPOL1 (NP_620059, 321 a.a. ~ 423 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 145282
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.