Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MIR16/GDE1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981891
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1891 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1891 Supplier Bioss Supplier No. BS12039R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Has glycerophosphoinositol phosphodiesterase activity. Has little or no activity towards glycerophosphocholine. GDE1 activity can be modulated by G-protein signaling pathways.

Specifications

Antigen MIR16/GDE1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Gde1
Gene Accession No. Q9NZC3
Gene Alias 1200003M13Rik; 363E6.2; BOS_22657; Gde1; Glycerophosphodiester phosphodiesterase 1; LOC100353916; membrane interacting protein of RGS16; Membrane-interacting protein of RGS16; Mir16; PAMCI; PCIP1; RGS16; RGS16-interacting membrane protein; similar to membrane interacting protein of RGS16; zgc:56068; zgc:56068 protein; zgc:77135
Gene Symbols Gde1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human MIR16/GDE1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51573
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.