Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MIR7-3HG Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA549100
Description
MIR7-3HG Polyclonal Antibody for Western Blot

Specifications
MIR7-3HG | |
Polyclonal | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q8N6C7 | |
MIR7-3HG | |
KLH conjugated synthetic peptide between 26-54 amino acids from the Central region of human PGSF1. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
MIR7-3HG | |
C19orf30; Huh7; LINC00306; MIR7-3 host gene; MIR7-3HG; NCRNA00306; PGSF1; uc002mbe.2 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
284424 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction