Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Miraculin Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144502
Description
Miraculin Polyclonal Antibody for Western Blot, ELISA
Miraculin has the property of modifying a sour taste into a sweet taste. This alteration of taste perception persists for many minutes.
Specifications
Miraculin | |
Polyclonal | |
Unconjugated | |
Miraculin | |
Recombinant Synsepalum dulcificum Miraculin (30-220AA). | |
100 μL | |
Primary | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
4.33 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
Rabbit | |
Protein A/G | |
RUO | |
Plant | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction