Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mirin
GREENER_CHOICESDP

Catalog No. 319050 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
31-905-0 50 mg
31-901-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 31-905-0 Supplier R&D Systems™ Supplier No. 3190/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. MRN-ATM pathway inhibitor

Mre11-Rad50-Nbs1 (MRN)-ATM pathway inhibitor that blocks the 3' and 5' exonuclease activity associated with Mre11. Prevents ATM activation in response to double strand breaks (IC50 = 12 μM) and induces G2 cell cycle arrest. Also blocks homology-directed repair in vitro.

Chemical Identifiers

CAS 1198097-97-0
Molecular Formula C10H8N2O2S

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 220.25
Purity >99%
Molecular Weight (g/mol) 220.246
Chemical Name or Material Mirin
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.