Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mirus Bio™ TransIT-SiQUEST™ siRNA Transfection Reagent
Click to view available options
Quantity:
0.4 mL
1.5 mL
5 x 1.5 mL
10 x 1.5 mL
Description
- Broad Spectrum siRNA Delivery: Utilize one transfection reagent and protocol for a wide variety of cells
- Low Cellular Toxicity: Maintain cell density and reduce experimental biases
- Reproducible Results: Obtain consistent, targeted gene knockdowns from day to day
- High Knockdown Efficiency: Achieve optimal gene silencing in a large percentage of cells to ensure experimental success
Specifications
Specifications
Content And Storage | 4°C |
Cell Type | Mammalian |
Format | Tube |
Promoter | None (Promoterless) |
Transfection Technique | Chemical Transfection |
For Use With (Application) | Enables high efficiency siRNA delivery and knockdown of target gene expression in many cell types including primary cells |
Includes | Ethanol and Water |
Product Type | Transfection Reagent |
Quantity | 0.4 mL |
Sterility | Sterile |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction