Learn More
Description
- Sensitive - Easily visualize transfected cells and assess transfection efficiency using fluorescent microscopy
- Inert - Does not target any mammalian genes or cause off-target effects
- Compatible - Suitable for co-delivery experiments with functional siRNA
- Ease of Use - Supplied as a ready to use 10 μM stock solution with a 10X RNAi Dilution Buffer
- Visualization and tracking of transfected RNAi duplexes
Specifications
Specifications
| Format | Liquid |
| Product Type | Label IT RNAi Delivery Control |
| Stability | 6 Months |
| Storage Buffer | Dilution Buffer (50 mM Tris, pH 7.5, 100 mM NaCl) |
| Content And Storage | -20°C |
| Concentration | 0.75 nmol |
| Label or Dye | Cy™3 |
| Quantity | 10 μg |
| For Use With (Application) | in-vitro or in vivo tracking using direct fluorescent microscopy, co-delivery experiments with functional siRNA |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.