Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio™ Label IT™ RNAi Delivery Controls

Catalog No. MIR7903 Shop All Mirus Bio LLC Products
Change view
Click to view available options
Quantity:
10 μg
100 μg
Label or Dye:
Cy™3
Fluorescein
Concentration:
0.75 nmol
7.5 nmol
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Label or Dye Concentration
MIR-7903 100 μg Fluorescein 7.5 nmol
MIR-7901 100 μg Cy™3 7.5 nmol
MIR-7900 10 μg Cy™3 0.75 nmol
MIR-7902 10 μg Fluorescein 0.75 nmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MIR-7903 Supplier Mirus Bio™ Supplier No. MIR 7903

Label IT™ RNAi Delivery Controls feature fluorescently labeled RNAi duplexes ideal for In Vitro or In Vivo tracking using direct fluorescent microscopy.

  • Sensitive - Easily visualize transfected cells and assess transfection efficiency using fluorescent microscopy
  • Inert - Does not target any mammalian genes or cause off-target effects
  • Compatible - Suitable for co-delivery experiments with functional siRNA
  • Ease of Use - Supplied as a ready to use 10 μM stock solution with a 10X RNAi Dilution Buffer
  • Visualization and tracking of transfected RNAi duplexes

Specifications

Format Liquid
Product Type Label IT RNAi Delivery Control
Stability 6 Months
Storage Buffer Dilution Buffer (50 mM Tris, pH 7.5, 100 mM NaCl)
Content And Storage -20°C
Concentration 7.5 nmol
Label or Dye Fluorescein
Quantity 100 μg
Product Line Molecular Probes, Click-iT
For Use With (Application) in-vitro or in vivo tracking using direct fluorescent microscopy, co-delivery experiments with functional siRNA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.