Learn More
Description
- Broad spectrum DNA delivery: Achieve high expression levels in breast cancer cell types including MCF-7, MDA-MB-231, MDA-MB-453, MDA-MB-468 and T47D
- Formulated for low cellular toxicity: Maintain cell density and reduce experimental biases due to toxicity
- Multiple applications: Transient and stable transfections for gene expression, shRNA expression and promoter analyses
Specifications
Specifications
| Cell Type | Cancer Cells |
| Format | Tube |
| Promoter | None (Promoterless) |
| Transfection Technique | Chemical Transfection |
| For Use With (Application) | Achieve high expression levels in breast cancer cell types including MCF-7, MDA-MB-231, MDA-MB-453, MDA-MB-468 and T47D, transient and stable transfections for gene expression, shRNA expression and promoter analyses |
| Product Type | Transfection Reagent |
| Quantity | 5 x 1 mL |
| Sterility | Sterile |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.