Learn More
Description
Includes
TransIT-CHO reagent and CHO Mojo reagent, both supplied as ready-to-use solutions.
- TransIT-CHO reagent and CHO Mojo reagent, both supplied as ready-to-use solutions.
- Unique Formulation: Each kit contains a base reagent and a booster reagent to optimize transfection performance in CHO cells
- Low Cellular Toxicity: Maintains cell density and reduces experimental bias
- High Efficiency Delivery: Allows 50 to 60% transfection efficiency in CHO-K1 cells
- Suitable for both transient and stable transfection
Specifications
Specifications
| Content And Storage | Store at -20°C |
| Cell Type | CHO |
| Format | Tube |
| Promoter | None (Promoterless) |
| Transfection Technique | Chemical Transfection |
| For Use With (Application) | Specifically optimized to provide exceptional transfection efficiency of plasmid DNA in CHO cells and cell types of associated lineage |
| Includes | 0.4 mL 2.22 mg/mL in 80% Ethanol, TransIT™-CHO reagent and 0.28 mL 5 mg/mL in 100% CHO Mojo reagent, both supplied as ready-to-use solution |
| Product Type | Transfection Reagent |
| Quantity | 0.4 mL |
| Sterility | Sterile |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.