Learn More
Description
- Transient transfection and recombinant baculovirus production are commonly used methods for insect cell expression
- Animal-origin free transfection reagent specifically optimized for high gene expression in a variety of insect cell types
- Exceptional DNA delivery in insect cell types including Sf9, High Five™ and S2
- High baculovirus production is ideal for baculovirus expression in insect cells
- Serum compatibility is non-liposomal, animal-origin free formulation that eliminates media change
- Low reagent amounts required per transfection
Specifications
Specifications
| Content And Storage | -20°C |
| Cell Type | Insect Cells |
| Format | Tube |
| Promoter | None (Promoterless) |
| Transfection Technique | Chemical Transfection |
| Species | Insect, Drosophila |
| For Use With (Application) | Ideal for baculovirus expression in insect cells |
| Product Type | Transfection Reagent |
| Quantity | 1.0 mL |
| Sterility | Sterile |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.