Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio™ TransIT™-Insect Transfection Reagent

Catalog No. p-4692983
Click to view available options
Quantity:
0.4 mL
1.0 mL
5 x 1.0 mL
10 x 1.0 mL

Use TransIT™-Insect Transfection Reagent to produce proteins with simple post-translational modifications. Low reagent amounts required per transfection.

  • Transient transfection and recombinant baculovirus production are commonly used methods for insect cell expression
  • Animal-origin free transfection reagent specifically optimized for high gene expression in a variety of insect cell types
  • Exceptional DNA delivery in insect cell types including Sf9, High Five™ and S2
  • High baculovirus production is ideal for baculovirus expression in insect cells
  • Serum compatibility is non-liposomal, animal-origin free formulation that eliminates media change
  • Low reagent amounts required per transfection

Specifications

Content And Storage -20°C
Cell Type Insect Cells
Format Tube
Promoter None (Promoterless)
Transfection Technique Chemical Transfection
Species Insect, Drosophila
For Use With (Application) Ideal for baculovirus expression in insect cells
Product Type Transfection Reagent
Quantity 0.4 mL
Sterility Sterile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.