Learn More
Description
Includes
TransIT-Keratinocyte transfection reagent, supplied as a ready-to-use solution.
- TransIT-Keratinocyte transfection reagent, supplied as a ready-to-use solution.
- Optimized transfection performance in keratinocyte cells.
- Low cellular toxicity maintains cell density and reduces experimental bias.
- Allows 20 to 30% transfection efficiency in keratinocytes.
- Concentration: 1.37 mg/mL
Specifications
Specifications
| Cell Type | Mammalian |
| Format | Tube |
| Promoter | None (Promoterless) |
| Transfection Technique | Chemical Transfection |
| Concentration | 1.37 mg/mL |
| For Use With (Application) | Highly efficient DNA or RNA transfection with minimal cellular toxicity, specifically for primary keratinocytes |
| Product Type | Transfection Reagent |
| Quantity | 1 mL |
| Sterility | Sterile |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.