Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio™ TransIT™-mRNA Transfection Kit
GSA_VA

Catalog No. MIR2250 Shop All Mirus Bio LLC Products
Change view
Click to view available options
Quantity:
0.4 mL
1 mL
5 mL
10 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MIR-2250 1 mL
MIR-2255 5 mL
MIR-2225 0.4 mL
MIR2256 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MIR-2250 Supplier Mirus Bio™ Supplier No. MIR 2250
Add to Cart
Add to Cart

Includes

TransIT-mRNA Transfection Reagent; mRNA Boost Reagent

Developed specifically for high efficiency delivery of both mRNA and viral RNA to various types of mammalian cells. Optimized for use in presence of serum, saving assay time and eliminating media changes. Transcriptional regulation effects are avoided by directly delivering the RNA to the cytoplasm.

  • Low Cellular Toxicity: Maintain cell density and reduce experimental biases
  • High Efficiency Delivery: Achieve RNA delivery in large population of cells to ensure experimental success
  • Serum Compatible: Perform transfections in presence of serum which eliminates need for media change and maintains cellular health
  • Deliver Various Sizes of RNA: Ideal for specialized applications, such as viral production and protein expression from mRNA
High efficiency delivery of both mRNA and viral RNA

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Cell Type Mammalian
Format Tube
Promoter None (Promoterless)
Transfection Technique Chemical Transfection
For Use With (Application) High efficiency delivery of both mRNA and viral RNA to various types of mammalian cells
Includes 1 mL TransIT™-mRNA Transfection Reagent, 1 mL mRNA Boost Reagent
Product Type Transfection Reagent
Quantity 1 mL
Sterility Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.