Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio™ TransIT™-Oligo Transfection Reagent

Catalog No. p-319690
Click to view available options
Quantity:
1 mL
5 x 1 mL
10 x 1 mL

TransIT™-Oligo Transfection Reagent provides efficient transfection of a wide range of oligonucleotides and oligoribonucleotides into a variety of cell types. Transfections do not require media changes and can be performed in serum-containing media.

  • Unique Formulation: Maximize transfection performance of oligonucleotides into a wide range of cells
  • Low Cellular Toxicity: Maintain cell density and reduce experimental biases
  • High Efficiency Delivery: Achieve high transfection efficiency in cells to ensure experimental success
  • One milliliter is sufficient to perform up to 500 transfections in 24-well plates
  • Tested on a variety of cell lines and oligonucleotides

Specifications

Cell Type Mammalian
Format Tube
Promoter None (Promoterless)
Transfection Technique Chemical Transfection
For Use With (Application) Provides efficient transfection of a wide range of Oligonucleotides and Oligoribonucleotides into a variety of cell types
Product Type Transfection Reagent
Quantity 1 mL
Sterility Sterile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.