Learn More
Description
- Broad Spectrum siRNA Delivery: Utilize one transfection reagent and protocol for a wide variety of cells
- Low Cellular Toxicity: Maintain cell density and reduce experimental biases
- Reproducible Results: Obtain consistent, targeted gene knockdowns from day to day
- High Knockdown Efficiency: Achieve optimal gene silencing in a large percentage of cells to ensure experimental success
- Concentration: 1.37 mg/mL
Specifications
Specifications
| Cell Type | Mammalian |
| Format | Tube |
| Promoter | None (Promoterless) |
| Transfection Technique | Chemical Transfection |
| Concentration | 1.37 mg/mL |
| For Use With (Application) | Transfect small interfering RNA for knock out of target gene expression, optimal gene silencing in a large percentage of cells |
| Product Type | Transfection Reagent |
| Quantity | 5 x 1.5 mL |
| Sterility | Sterile |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.