Learn More
Description
- Efficiency: Provides exceptional broad spectrum transfection
- Versatility: Facilitates cutting edge delivery of plasmid DNA, siRNA and miRNA
- Functions on a novel, non-liposomal polymeric technology
Specifications
Specifications
| Content And Storage | -20°C |
| Cell Type | Established Cell Lines, Stem Cells, Primary Cells, Hard-to-Transfect Cells |
| Format | Tube |
| Promoter | None (Promoterless) |
| Transfection Technique | Chemical Transfection |
| For Use With (Application) | High efficiency delivery of plasmid DNA and siRNA/miRNA |
| Product Type | Transfection Reagent |
| Quantity | 5 x 1.5 mL |
| Sterility | Sterile |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.