Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio™ TransIT-X2™ Dynamic Delivery System

Catalog No. MIR6006 Shop All Mirus Bio LLC Products
Change view
Click to view available options
Quantity:
0.3 mL
0.75 mL
1.5 mL
5 x 1.5 mL
10 x 1.5 mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MIR6006 10 x 1.5 mL
MIR6003 0.3 mL
MIR6004 0.75 mL
MIR6000 1.5 mL
MIR6005 5 x 1.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MIR6006 Supplier Mirus Bio™ Supplier No. MIR6006
Add to Cart
Add to Cart
Request a Quote

Non-liposomal polymeric system for efficient transfection of plasmid DNA and siRNA/miRNA. Transfections do not require medium changes and can be performed in serum-containing medium.

  • Efficiency: Provides exceptional broad spectrum transfection
  • Versatility: Facilitates cutting edge delivery of plasmid DNA, siRNA and miRNA
  • Functions on a novel, non-liposomal polymeric technology

Specifications

Content And Storage -20°C
Cell Type Established Cell Lines, Stem Cells, Primary Cells, Hard-to-Transfect Cells
Format Tube
Promoter None (Promoterless)
Transfection Technique Chemical Transfection
For Use With (Application) High efficiency delivery of plasmid DNA and siRNA/miRNA
Product Type Transfection Reagent
Quantity 10 x 1.5 mL
Sterility Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.