Learn More
Description
- MIR 5910 provides sufficient reagents for three endotoxin extraction rounds on a total of at least 10 mg of DNA
- MIR 5900 provides sufficient reagents for three endotoxin extraction rounds on a total of at least 100 mg of DNA
- Effectively Removes Endotoxin from Plasmid Samples: Aids in the safety and efficiency of gene delivery research
- Compatible with In Vivo and In Vitro Applications: Removes harmful endotoxin that could cause decreased transfection efficiencies in vitro and inflammatory reactions in vivo
- Ease of Use Simple phase separation protocol allows quick visualization of recovered nucleic acid preparations
Specifications
Specifications
| Content And Storage | 4°C |
| Format | Kit |
| Isolation Technology | Organic Extraction |
| For Use With (Application) | Removes bacterial endotoxins from Nucleic Acids for both in vivo and in-vitro applications |
| Product Type | Endotoxin Removal Kit |
| Quantity | For 100 mg DNA |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.