Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MIS/AMH Antibody (003), mFluor Violet 610 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290330MFV610
Description
MIS/AMH Monoclonal antibody specifically detects MIS/AMH in Human samples. It is validated for ELISA, ELISASpecifications
MIS/AMH | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
268 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
003 | |
ELISA, Sandwich ELISA Detection | |
MIF, MIS, Muellerian hormone, muellerian-inhibiting factor, muellerian-inhibiting substance, Mullerian hormone, Mullerian inhibiting factor, Mullerian inhibiting substance | |
Rabbit immunized with purified, recombinant Human MIS/AMH (Accession#: NP_000470.2, Leu25-Arg560) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction