Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MIS/AMH Antibody (AMH/6713R), DyLight 594, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP308459DL594
Description
MIS/AMH Monoclonal antibody specifically detects MIS/AMH in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
MIS/AMH | |
Monoclonal | |
DyLight 594 | |
MIF, MIS, Muellerian hormone, muellerian-inhibiting factor, muellerian-inhibiting substance, Mullerian hormone, Mullerian inhibiting factor, Mullerian inhibiting substance | |
A synthetic peptide from the C-terminus of human MIS/AMH (aa 460-560) (exact sequence is proprietary) (Uniprot: IDP03971) | |
100 μg | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
AMH/6713R | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified | |
RUO | |
268 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction