Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MIS/AMH Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP243670
Description
Specifically detects MIS/AMH in Human, Mouse samples, and it is validated for Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blotting
MIS/AMH Polyclonal antibody specifically detects MIS/AMH in Human, Mouse, Feline samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).Specifications
MIS/AMH | |
Polyclonal | |
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
MIF, MIS, Muellerian hormone, muellerian-inhibiting factor, muellerian-inhibiting substance, Mullerian hormone, Mullerian inhibiting factor, Mullerian inhibiting substance | |
Rabbit | |
59 kDa | |
0.1 mL | |
Primary | |
Human, Mouse, Feline | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.0) and 20% Glycerol with 0.025% Proclin 300 | |
AMH | |
Recombinant protein encompassing a sequence within the C-terminus region of human AMH. The exact sequence is proprietary. | |
Immunogen affinity purified | |
RUO | |
268 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction