Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MIS/AMH Antibody (RM0288-8D29), Alexa Fluor™ 532, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP211801AF532
Description
MIS/AMH Monoclonal specifically detects MIS/AMH in Mouse samples. It is validated for Western Blot.Specifications
MIS/AMH | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
AMH | |
Mouse recombinant MIS/AMH | |
0.1 mL | |
268.0 | |
Store at 4°C in the dark. |
Western Blot | |
RM0288-8D29 | |
Western Blot | |
MIF, MIS, Muellerian hormone, muellerian-inhibiting factor, muellerian-inhibiting substance, Mullerian hormone, Mullerian inhibiting factor, Mullerian inhibiting substance | |
Rat | |
Protein A or G purified | |
Primary | |
Mouse | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction