Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MIS/AMH Goat anti-Mouse, Rat, Polyclonal, R&D Systems™

Catalog No. AF1446SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1446SP 25 μg
AF1446 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF1446SP Supplier R&D Systems Supplier No. AF1446SP
This item is not returnable. View return policy

Goat Polyclonal Antibody

MIS/AMH Polyclonal antibody AF1446SP Specifically detects MIS/AMH in Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blotting

Specifications

Antigen MIS/AMH
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P49000
Gene Alias AMH, MIF, MIS, Muellerian hormone, muellerian-inhibiting factor, muellerian-inhibiting substance, Mullerian hormone, Mullerian inhibiting factor, Mullerian inhibiting substance
Gene Symbols AMH
Host Species Goat
Immunogen E. coli-derived recombinant rat MIS/AMH Lys447-Arg553 Accession # P49000
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 268
Test Specificity Detects mouse and rat MIS/AMH in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant human MIS is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.