Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MIS RII/AMHR2 Antibody (SR1978), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322158100UL
Description
MIS RII/AMHR2 Monoclonal antibody specifically detects MIS RII/AMHR2 in Human, Mouse samples. It is validated for Western BlotSpecifications
MIS RII/AMHR2 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot | |
SR1978 | |
Western Blot 1:500-1:2000 | |
AMH type II receptor, AMHREC 2.7.11.30, MISR2, MISRIIMIS type II receptor, MRII, Muellerian hormone type II receptor, Muellerian hormone type-2 receptor, Mullerian hormone receptor, type II, Mullerian inhibiting substance type II receptor | |
A synthesized peptide derived from human MIS RII/AMHR2 (Uniprot #: Q16671) | |
100 μg | |
Protein Kinase | |
269 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction