Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MIS12 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1002359AF350
Description
MIS12 Polyclonal antibody specifically detects MIS12 in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
MIS12 | |
Polyclonal | |
Western Blot, Immunohistochemistry | |
2510025F08Rik, hMis12, homolog of yeast Mis12, KNTC2AP, MGC2488, MIS12 homolog, MIS12, MIND kinetochore complex component, homolog (S. pombe), MIS12, MIND kinetochore complex component, homolog (yeast), MTW1, protein MIS12 homolog | |
The immunogen recognized by this antibody maps to a region between residue 150 and the C-terminus (residue 205) of human MIS12 homolog using the numbering given in entry NP_076944.1 (GeneID 79003). | |
0.1 mL | |
Cell Cycle and Replication, DNA Repair, modENCODE Antibodies | |
79003 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction