Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MIS12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5115577
Description
Antibody detects endogenous levels of total MIS12.
This protein is part of the MIS12 complex which is required for normal chromosome alignment and segregation and for kinetochore formation during mitosis.
Specifications
MIS12 | |
Polyclonal | |
Unconjugated | |
MIS12 | |
2510025F08Rik; Ac1585; AU044651; hMis12; homolog of yeast Mis12; KNTC2AP; Liver regeneration-related protein LRRG112; Mis12; MIS12 homolog; MIS12 homolog (yeast); MIS12 kinetochore complex component; MIS12, kinetochore complex component; MIS12, MIND kinetochore complex component, homolog; Misc12; MTW1; Protein MIS12 homolog | |
Rabbit | |
Affinity chromatography | |
RUO | |
501706, 67139, 79003 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
Q7TQ72, Q9CY25, Q9H081 | |
MIS12 | |
A synthesized peptide derived from human MIS12(Accession Q9H081), corresponding to amino acid residues E83-C133. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction