Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MIS12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA540125
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA540125 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA540125 Supplier Invitrogen™ Supplier No. PA540125
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HeLa and HepG2 cells. Predicted reactivity: Rhesus Monkey (97%), Bovine (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This protein is part of the MIS12 complex which is required for normal chromosome alignment and segregation and for kinetochore formation during mitosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MIS12
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.99 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene MIS12
Gene Accession No. Q9H081
Gene Alias 2510025F08Rik; Ac1585; AU044651; hMis12; homolog of yeast Mis12; KNTC2AP; Liver regeneration-related protein LRRG112; Mis12; MIS12 homolog; MIS12 homolog (yeast); MIS12 kinetochore complex component; MIS12, kinetochore complex component; MIS12, MIND kinetochore complex component, homolog; Misc12; MTW1; Protein MIS12 homolog
Gene Symbols MIS12
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human MIS12. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79003
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.