Learn More
MIS12 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300775AT
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Bovine and Orangutan
This protein is part of the MIS12 complex which is required for normal chromosome alignment and segregation and for kinetochore formation during mitosis.Specifications
| MIS12 | |
| Polyclonal | |
| Unconjugated | |
| MIS12 | |
| MTW1, hMis12, 2510025F08Rik, KNTC2AP, MIS12, MIND kinetochore complex component, homolog (S. pombe) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79003 | |
| 4° C | |
| Liquid |
| Immunoprecipitation | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Q9H081 | |
| MIS12 | |
| Between 1 and 50 | |
| 10 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.