Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MIS18A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554238
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554238 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554238 Supplier Invitrogen™ Supplier No. PA554238
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAGVRSLRCS RGCAGGCECG DKGKCSDSSL LGKRLSEDSS RHQLLQKWAS MWSSMSEDAS VADMERAQLE EEAAAAEERP LVF Highest antigen sequence identity to the following orthologs: Mouse - 41%, Rat - 57%.

C21orf45, also named as MIS18A, C21orf46 and FASP1, is required for recruitment of CENPA to centromeres and normal chromosome segregation during mitosis. C21orf45 has dimer (~52 kDa) and heterodimer(~105 kDa) forms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MIS18A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MIS18A
Gene Accession No. Q9NYP9
Gene Alias 2610039C10Rik; 2810018N07Rik; B28; C21orf45; C21orf46; FAPP1-associated protein 1; FASP1; hMis18alpha; MIS18 kinetochore protein A; MIS18 kinetochore protein homolog A; MIS18 kinetochore protein homolog A (S. pombe); Mis18a; MIS18alpha; protein Mis18-alpha; RGD1310778
Gene Symbols MIS18A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MIS18A. Recombinant protein control fragment (Product #RP-91767).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54069
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.