Learn More
Invitrogen™ MIS18BP1 Polyclonal Antibody

Description
Immunogen sequence: QHQRILLPSF QDSEDDDDIL PNMDKNPTTP SSVIFPLVKT PQCQHVSPGM LGSINRNDCD KYVFRMQKYH KSNGGIVWGN IKKKLVETDF STPTPRRKTP FNTDLGENSG IGKLFTNAVE SLDE Highest antigen sequence identity to the following orthologs: Mouse - 57%, Rat - 61%.
Specifications
Specifications
| Antigen | MIS18BP1 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | MIS18BP1 |
| Gene Accession No. | Q6P0N0 |
| Gene Alias | 6720403H11; C14orf106; C79407; HSA242977; HsKNL-2; KIAA1903; kinetochore null 2 homolog; kinetochore-associated protein KNL-2 homolog; Knl2; M18BP1; MIS18 binding protein 1; mis18-binding protein 1; Mis18bp1; mKIAA1903; P243; putative protein p243 which interacts with transcription factor Sp1; RGD1305497 |
| Gene Symbols | MIS18BP1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.