Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MIS18BP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA546777
Description
Peptide sequence: ECQRKYMENP RGKGSQKHVT KKKPANSKGQ NGKRGDADQK QTIKITAKVG.
MIS18BP1 is required for recruitment of CENPA to centromeres and normal chromosome segregation during mitosis.
Specifications
| MIS18BP1 | |
| Polyclonal | |
| Unconjugated | |
| MIS18BP1 | |
| 6720403H11; C14orf106; C79407; HSA242977; HsKNL-2; KIAA1903; kinetochore null 2 homolog; kinetochore-associated protein KNL-2 homolog; Knl2; M18BP1; MIS18 binding protein 1; mis18-binding protein 1; Mis18bp1; mKIAA1903; P243; putative protein p243 which interacts with transcription factor Sp1; RGD1305497 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 55320 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q6P0N0 | |
| MIS18BP1 | |
| Synthetic peptide directed towards the C-terminal of human MIS18BP1 (aa 917-966). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction