Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MIST1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP282008
Description
MIST1 Polyclonal specifically detects MIST1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.Specifications
MIST1 | |
Polyclonal | |
Unconjugated | |
PBS | |
BHLHA15 | |
MIST1 antibody was raised against a peptide corresponding to 17 amino acids near the carboxy terminus of human MIST1. Amino Acid Squence: ATEAQPQGHLQRYSTQI | |
Affinity Purified | |
RUO | |
168620 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot, ELISA | |
1 mg/mL | |
Western Blot 1-2 μg/mL, ELISA | |
basic helix-loop-helix domain containing, class B, 8, basic helix-loop-helix family, member a15, bHLHa15, bHLHb8, class A basic helix-loop-helix protein 15, Class B basic helix-loop-helix protein 8, class II bHLH protein MIST1, MIST1MIST-1, Muscle, intestine and stomach expression 1 | |
Rabbit | |
22 kDa | |
0.1 mg | |
Primary | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction