Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Mist1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583883
Description
Immunogen sequence: AKNYIKSLTA TILTMSSSRL PGLEGPGPKL YQHYQQQQQV AGGALGATEA QPQGHLQRYS TQIHSFREG.
Plays a role in controlling the transcriptional activity of MYOD1, ensuring that expanding myoblast populations remain undifferentiated. Repression may occur through muscle-specific E-box occupancy by homodimers. May also negatively regulate bHLH-mediated transcription through an N-terminal repressor domain. Serves as a key regulator of acinar cell function, stability, and identity. Also required for normal organelle localization in exocrine cells and for mitochondrial calcium ion transport. May function as a unique regulator of gene expression in several different embryonic and postnatal cell lineages. Binds to the E-box consensus sequence 5'-CANNTG-3'.
Specifications
Mist1 | |
Polyclonal | |
Unconjugated | |
Bhlha15 | |
1810009C13Rik; basic helix-loop-helix domain containing, class B, 8; basic helix-loop-helix family member a15; basic helix-loop-helix family, member a15; Basic helix-loop-helix transcription factor (bHlH); bHlH; BHLHA15; BHLHB8; Class A basic helix-loop-helix protein 15; class B basic helix-loop-helix protein 8; class II bHLH protein MIST1; MIST1; MIST-1; Muscle, intestine and stomach expression 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
168620 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.08 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7RTS1 | |
Bhlha15 | |
Recombinant protein corresponding to Human BHLHA15. Recombinant protein control fragment (Product #RP-99943). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction