Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mist1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114515
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114515 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114515 Supplier Invitrogen™ Supplier No. PA5114515
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive controls that may be used include: 3T3 (NIH) Cell Lysate.

Plays a role in controlling the transcriptional activity of MYOD1, ensuring that expanding myoblast populations remain undifferentiated. Repression may occur through muscle-specific E-box occupancy by homodimers. May also negatively regulate bHLH-mediated transcription through an N-terminal repressor domain. Serves as a key regulator of acinar cell function, stability, and identity. Also required for normal organelle localization in exocrine cells and for mitochondrial calcium ion transport. May function as a unique regulator of gene expression in several different embryonic and postnatal cell lineages. Binds to the E-box consensus sequence 5'-CANNTG-3'.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mist1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Bhlha15
Gene Accession No. P70562, Q7RTS1, Q9QYC3
Gene Alias 1810009C13Rik; basic helix-loop-helix domain containing, class B, 8; basic helix-loop-helix family member a15; basic helix-loop-helix family, member a15; Basic helix-loop-helix transcription factor (bHlH); bHlH; BHLHA15; BHLHB8; Class A basic helix-loop-helix protein 15; class B basic helix-loop-helix protein 8; class II bHLH protein MIST1; MIST1; MIST-1; Muscle, intestine and stomach expression 1
Gene Symbols Bhlha15
Host Species Rabbit
Immunogen BHLHA15 antibody was raised against a peptide corresponding to 17 amino acids near the carboxy terminus of human BHLHA15.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 168620, 17341, 25334
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.