Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Mist1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5114474
Description
Positive controls that may be used include: Rat Small Intestine Tissue Lysate, Human Small Intestine Tissue Slide.
Plays a role in controlling the transcriptional activity of MYOD1, ensuring that expanding myoblast populations remain undifferentiated. Repression may occur through muscle-specific E-box occupancy by homodimers. May also negatively regulate bHLH-mediated transcription through an N-terminal repressor domain. Serves as a key regulator of acinar cell function, stability, and identity. Also required for normal organelle localization in exocrine cells and for mitochondrial calcium ion transport. May function as a unique regulator of gene expression in several different embryonic and postnatal cell lineages. Binds to the E-box consensus sequence 5'-CANNTG-3'.
Specifications
Mist1 | |
Polyclonal | |
Unconjugated | |
Bhlha15 | |
1810009C13Rik; basic helix-loop-helix domain containing, class B, 8; basic helix-loop-helix family member a15; basic helix-loop-helix family, member a15; Basic helix-loop-helix transcription factor (bHlH); bHlH; BHLHA15; BHLHB8; Class A basic helix-loop-helix protein 15; class B basic helix-loop-helix protein 8; class II bHLH protein MIST1; MIST1; MIST-1; Muscle, intestine and stomach expression 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
168620, 17341, 25334 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
P70562, Q7RTS1, Q9QYC3 | |
Bhlha15 | |
BHLHA15 antibody was raised against a 18 amino acid peptide near the center of human BHLHA15. The immunogen is located within amino acids 60 - 110 of BHLHA15. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction