Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mitchell Plastics Biohazard Floor Waste Containers
Click to view available options
Height (English):
29 in.
30.25 in.
Length (English):
14 in.
17.25 in.
Width (English):
14 in.
17.25 in.
For Use With (Equipment):
30.48 x 30.48 x 68.58 cm Box
91.44 x 114.3 cm Bag
Description
- Made of heavy-duty acrylic to help protect personnel from exposure to potential hazards
- Clear design enables warning insignias and biohazard bag colors to be seen
- Each unit has a hinged lid for easy placement of waste
- Lids are also removable for easy positioning and removal of bag or box
- Reusable
Floor models
- Feature removable bottoms and can be used for radioactive material disposal
- WC-100: holds bag 91.44 x 114.3cm (36 x 45 in.)
- WB-500: holds box 30.48 x 30.48 x 68.58cm (12 x 12 x 27 in.)
Specifications
Specifications
| Color | Clear |
| Dimensions (L x W x H) | 17.25 x 17.25 x 30.25 in. (43.84 x 44.5 x 76.84 cm) |
| Height (Metric) | 76.84 cm |
| Length (Metric) | 43.84 cm |
| Model | Floor |
| Size | Large |
| Width (Metric) | 44.5 cm |
| For Use With (Application) | For radioactive material |
| For Use With (Equipment) | 91.44 x 114.3 cm Bag |
| Width (English) | 17.25 in. |
| Show More |
Dispose contents according to all local, state and federal disposal regulations.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction