Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MITD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557549
Description
Immunogen sequence: MAKSGLRQDP QSTAAATVLK RAVELDSESR YPQALVCYQE GIDLLLQVLK GTKDNTKRCN LREKISKYMD RAENIKKYLD Q Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 80%.
MITD1 may play a role in endosomal protein transport.
Specifications
MITD1 | |
Polyclonal | |
Unconjugated | |
MITD1 | |
1500032H18Rik; hypothetical protein MGC56159; microtubule interacting and trafficking domain containing 1; MIT domain-containing protein 1; MIT, microtubule interacting and transport, domain containing 1; Mitd1; RGD1307700; zgc:56159 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
129531 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WV92 | |
MITD1 | |
Recombinant protein corresponding to Human MITD1. Recombinant protein control fragment (Product #RP-96161). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction