Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MITD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116854
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116854 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116854 Supplier Invitrogen™ Supplier No. PA5116854
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: U-87MG, MCF7, Mouse thymus, Mouse spleen, Rat thymus, Rat spleen Immunogen sequence: MAKSGLRQDP QSTAAATVLK RAVELDSESR YPQALVCYQE GIDLLLQVLK GTKDNTKRCN LREKISKYMD RAENIKKYLD QEKEDGKYHK QIKIEENATG FSYESLFREY LNETVTEVWI EDPYIRHTHQ LYNFLRFCEM LIKRPCKVKT IHLLTSLDEG IEQVQQSRGL QEIEESLRSH GVLLEVQYSS SIHDREIRFN NGWMIKIGRG LDYFKKPQSR FSLGYCDFDL RPCHETTVDI FHKKHTKNI.

MITD1 may play a role in endosomal protein transport.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MITD1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.04 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene MITD1
Gene Accession No. Q5I0J5, Q8VDV8, Q8WV92
Gene Alias 1500032H18Rik; hypothetical protein MGC56159; microtubule interacting and trafficking domain containing 1; MIT domain-containing protein 1; MIT, microtubule interacting and transport, domain containing 1; Mitd1; RGD1307700; zgc:56159
Gene Symbols MITD1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-249 of human MITD1 (NP_6201531).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 129531, 363219, 69028
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.