Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MITF Antibody (PCRP-MITF-1D9), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314121B
This item is not returnable.
View return policy
Description
MITF Monoclonal antibody specifically detects MITF in Human samples. It is validated for Western Blot, Flow Cytometry, ImmunofluorescenceSpecifications
MITF | |
Monoclonal | |
Biotin | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
4286 | |
Store at 4°C in the dark. | |
Purified |
Western Blot, Flow Cytometry, Immunofluorescence | |
PCRP-MITF-1D9 | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
BHLHE32, bHLHe32MI, Class E basic helix-loop-helix protein 32, microphthalmia-associated transcription factor, Waardenburg syndrome, type 2A, WS2A | |
Recombinant full-length human MITF protein (Uniprot: O75030) | |
0.1 mL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction